Product Details
- SNP ID
-
rs34385858
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:19155910 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AAGCCTTAACTATGTCATTACTCCC[A/C]TTGCTTAGACTTATAGTTGAAATGG
- Phenotype
-
MIM: 608260
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNH8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79743471] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNH8
- Gene Name
- potassium voltage-gated channel subfamily H member 8
View Full Product Details