Product Details

SNP ID
rs665714
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:83200219 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTTCAGCTTCTCTTACTACTCTC[A/C]CTCAGTCTACTTTGGTTAACTCACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD42 PubMed Links

Gene Details

Gene
ANKRD42
Gene Name
ankyrin repeat domain 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300972.1 Intron NP_001287901.1
NM_001300973.1 Intron NP_001287902.1
NM_001300975.1 Intron NP_001287904.1
NM_001300976.1 Intron NP_001287905.1
NM_001300977.1 Intron NP_001287906.1
NM_182603.3 Intron NP_872409.2
Gene
LOC100506282
Gene Name
uncharacterized LOC100506282
There are no transcripts associated with this gene.

View Full Product Details