Product Details

SNP ID
rs1562792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57739890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATGCCCCTGACTCCAGGGGCAG[C/T]AGTCCTGAGGCCCAGTGAGGTGGGG
Phenotype
MIM: 602703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRC7 PubMed Links

Gene Details

Gene
DRC7
Gene Name
dynein regulatory complex subunit 7
There are no transcripts associated with this gene.

Gene
KATNB1
Gene Name
katanin regulatory subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005886.2 Intron NP_005877.2
XM_005255772.4 Intron XP_005255829.1
XM_006721121.3 Intron XP_006721184.1
XM_006721122.2 Intron XP_006721185.1
XM_006721123.3 Intron XP_006721186.1
XM_011522810.2 Intron XP_011521112.1
XM_017022860.1 Intron XP_016878349.1
XM_017022861.1 Intron XP_016878350.1
XM_017022862.1 Intron XP_016878351.1
XM_017022863.1 Intron XP_016878352.1
XM_017022864.1 Intron XP_016878353.1

View Full Product Details