Product Details
- SNP ID
-
rs10894799
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:134339014 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTGGATAAATGAAGCCGAGATACA[C/T]AGGCAACAGGAATCCTAGCAGAAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLB1L2
PubMed Links
Gene Details
- Gene
- GLB1L2
- Gene Name
- galactosidase beta 1 like 2
View Full Product Details