Product Details

SNP ID
rs28360537
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:56966734 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CATGAAAGTAAGCCAGCCACACTGT[T/C]TAATTTCCGAAAAGACAAAGGCTGG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BEND6 PubMed Links

Gene Details

Gene
BEND6
Gene Name
BEN domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318539.1 Intron NP_001305468.1
NM_152731.2 Intron NP_689944.2
XM_005248889.1 Intron XP_005248946.1
XM_011514346.2 Intron XP_011512648.1
XM_017010406.1 Intron XP_016865895.1
XM_017010407.1 Intron XP_016865896.1
XM_017010408.1 Intron XP_016865897.1

View Full Product Details