Product Details
- SNP ID
-
rs34616943
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:56958200 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGAGAAGGGAATTTTATACATCA[C/G]TCCTGCCCACTAATGGGTAGTAGGC
- Phenotype
-
MIM: 113810
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BEND6
PubMed Links
Gene Details
- Gene
- BEND6
- Gene Name
- BEN domain containing 6
- Gene
- DST
- Gene Name
- dystonin
There are no transcripts associated with this gene.
View Full Product Details