Product Details
- SNP ID
-
rs866066
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5450953 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTGGACACGGGTCCAAGTCCACCG[C/T]CAGCTGCTTGCTAGTAACATGACTT
- Phenotype
-
MIM: 605402
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD274
PubMed Links
Gene Details
- Gene
- CD274
- Gene Name
- CD274 molecule
View Full Product Details