Product Details
- SNP ID
-
rs35533417
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:109648966 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAAGATGTCATTCCATTGTGTT[C/T]TGGCATTGACCCTTGCTCTGGGAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PALM2
PubMed Links
Gene Details
- Gene
- PALM2
- Gene Name
- paralemmin 2
View Full Product Details