Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001103184.3 | 10758 | UTR 3 | NP_001096654.1 | ||
NM_001277313.1 | 10758 | UTR 3 | NP_001264242.1 | ||
NM_001277314.1 | 10758 | Intron | NP_001264243.1 | ||
XM_011521504.2 | 10758 | UTR 3 | XP_011519806.1 | ||
XM_011521505.2 | 10758 | Intron | XP_011519807.1 | ||
XM_011521506.2 | 10758 | UTR 3 | XP_011519808.1 | ||
XM_011521507.2 | 10758 | Intron | XP_011519809.1 | ||
XM_011521509.2 | 10758 | UTR 3 | XP_011519811.1 | ||
XM_011521511.2 | 10758 | UTR 3 | XP_011519813.1 | ||
XM_017022130.1 | 10758 | UTR 3 | XP_016877619.1 | ||
XM_017022131.1 | 10758 | Intron | XP_016877620.1 | ||
XM_017022132.1 | 10758 | UTR 3 | XP_016877621.1 | ||
XM_017022133.1 | 10758 | UTR 3 | XP_016877622.1 | ||
XM_017022134.1 | 10758 | UTR 3 | XP_016877623.1 | ||
XM_017022135.1 | 10758 | UTR 3 | XP_016877624.1 |