Product Details

SNP ID
rs17479454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27811209 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAGCAACCCAAAGGGAAACAGG[C/T]GAACGATTTGAACAAGTAGTTCACA
Phenotype
MIM: 609353 MIM: 611210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESCO2 PubMed Links

Gene Details

Gene
ESCO2
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017420.2 Intron NP_001017420.1
XM_011544421.2 Intron XP_011542723.1
XM_011544422.2 Intron XP_011542724.1
Gene
PBK
Gene Name
PDZ binding kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278945.1 Intron NP_001265874.1
NM_018492.3 Intron NP_060962.2
XM_006716368.2 Intron XP_006716431.1

View Full Product Details