Product Details

SNP ID
rs4806625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54925609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGTTTGAAGCCAGGAGTTTGAGAA[A/C]CTCGGCAACACGGCCACACAGTGCA
Phenotype
MIM: 604530 MIM: 609661
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCR1 PubMed Links

Gene Details

Gene
NCR1
Gene Name
natural cytotoxicity triggering receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145457.2 Intron NP_001138929.2
NM_001145458.2 Intron NP_001138930.2
NM_001242356.2 Intron NP_001229285.1
NM_001242357.2 Intron NP_001229286.1
NM_004829.6 Intron NP_004820.2
XM_011527528.2 Intron XP_011525830.1
XM_011527529.2 Intron XP_011525831.1
XM_011527530.2 Intron XP_011525832.1
Gene
NLRP7
Gene Name
NLR family pyrin domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127255.1 Intron NP_001120727.1
NM_139176.3 Intron NP_631915.2
NM_206828.3 Intron NP_996611.2
XM_006723075.3 Intron XP_006723138.1
XM_006723076.3 Intron XP_006723139.1
XM_011526596.2 Intron XP_011524898.1
XM_011526599.2 Intron XP_011524901.1
XM_011526601.2 Intron XP_011524903.1

View Full Product Details