Product Details
- SNP ID
-
rs9877400
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:191273650 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TACACCATGCCTCTGAGATTTAGAT[C/T]ACAACTGTTAACACCTAATAGCCTA
- Phenotype
-
MIM: 610280
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSTN
PubMed Links
Gene Details
- Gene
- OSTN
- Gene Name
- osteocrin
There are no transcripts associated with this gene.
- Gene
- UTS2B
- Gene Name
- urotensin 2B
View Full Product Details