Product Details

SNP ID
rs12457980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35371058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAAAGGTTAGGGTGAGAAAGGT[A/G]GTCTGGGGAGAAGAATTAGTGATGC
Phenotype
MIM: 609600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF396 PubMed Links

Gene Details

Gene
ZNF396
Gene Name
zinc finger protein 396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322286.1 Intron NP_001309215.1
NM_001322290.1 Intron NP_001309219.1
NM_001322291.1 Intron NP_001309220.1
NM_145756.2 Intron NP_665699.1
XM_006722432.3 Intron XP_006722495.1
XM_017025687.1 Intron XP_016881176.1
XM_017025688.1 Intron XP_016881177.1
XM_017025689.1 Intron XP_016881178.1

View Full Product Details