Product Details

SNP ID
rs10033058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68311690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAGTGATGATTATGTGAGCCAA[C/G]AGAATTGTTTAGAATAGTGCCAGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
YTHDC1 PubMed Links

Gene Details

Gene
YTHDC1
Gene Name
YTH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005265706.2 Intron XP_005265763.1
XM_005265707.2 Intron XP_005265764.1
XM_005265708.2 Intron XP_005265765.1
XM_017008832.1 Intron XP_016864321.1
XM_017008833.1 Intron XP_016864322.1

View Full Product Details