Product Details

SNP ID
rs7379278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178135814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGGGCCGGATTGTTTGACCTTC[A/G]TGAGGTCCTGGCTTTTTTGAGCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
N4BP3 PubMed Links

Gene Details

Gene
N4BP3
Gene Name
NEDD4 binding protein 3
There are no transcripts associated with this gene.

Gene
RMND5B
Gene Name
required for meiotic nuclear division 5 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288794.1 Intron NP_001275723.1
NM_001288795.1 Intron NP_001275724.1
NM_022762.4 Intron NP_073599.2
XM_005265969.3 Intron XP_005266026.1
XM_005265971.4 Intron XP_005266028.1
XM_005265972.3 Intron XP_005266029.1
XM_017009727.1 Intron XP_016865216.1
XM_017009728.1 Intron XP_016865217.1

View Full Product Details