Product Details

SNP ID
rs12175038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:72626904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGACATGTGGTATGAGGAAGAT[A/G]ATTCTGACTTGGTTGATTTGAGGAA
Phenotype
MIM: 607357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNQ5 PubMed Links

Gene Details

Gene
KCNQ5
Gene Name
potassium voltage-gated channel subfamily Q member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160130.1 Intron NP_001153602.1
NM_001160132.1 Intron NP_001153604.1
NM_001160133.1 Intron NP_001153605.1
NM_001160134.1 Intron NP_001153606.1
NM_019842.3 Intron NP_062816.2
XM_017011058.1 Intron XP_016866547.1
Gene
KCNQ5-IT1
Gene Name
KCNQ5 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details