Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_023012.5 | 1861 | UTR 3 | NP_075388.2 | ||
XM_005253601.1 | 1861 | UTR 3 | XP_005253658.1 | ||
XM_005253602.2 | 1861 | UTR 3 | XP_005253659.1 | ||
XM_005253604.1 | 1861 | UTR 3 | XP_005253661.1 | ||
XM_011538688.1 | 1861 | UTR 3 | XP_011536990.1 | ||
XM_011538689.1 | 1861 | UTR 3 | XP_011536991.1 | ||
XM_017019830.1 | 1861 | UTR 3 | XP_016875319.1 | ||
XM_017019831.1 | 1861 | UTR 3 | XP_016875320.1 | ||
XM_017019832.1 | 1861 | UTR 3 | XP_016875321.1 | ||
XM_017019833.1 | 1861 | UTR 3 | XP_016875322.1 |