Product Details

SNP ID
rs10773230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122508464 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCTGCAGCTGCTTGAAGAGAATA[C/T]AAAAATGGATTTTAAAACGATTTTA
Phenotype
MIM: 616381
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSRC2 PubMed Links

Gene Details

Gene
RSRC2
Gene Name
arginine and serine rich coiled-coil 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023012.5 Intron NP_075388.2
XM_005253601.1 Intron XP_005253658.1
XM_005253602.2 Intron XP_005253659.1
XM_005253604.1 Intron XP_005253661.1
XM_011538688.1 Intron XP_011536990.1
XM_011538689.1 Intron XP_011536991.1
XM_017019830.1 Intron XP_016875319.1
XM_017019831.1 Intron XP_016875320.1
XM_017019832.1 Intron XP_016875321.1
XM_017019833.1 Intron XP_016875322.1
Gene
ZCCHC8
Gene Name
zinc finger CCHC-type containing 8
There are no transcripts associated with this gene.

View Full Product Details