Product Details

SNP ID
rs12223229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57545996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCTGCTCGGCCCCGGGGGCCC[C/G]GGGCACAGAACCGCAAAGCCATCGT
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105565.2 1035 Silent Mutation CGG,GGG R345G NP_001099035.2
XM_005273819.3 1035 Silent Mutation CGG,GGG R345G XP_005273876.1
Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
There are no transcripts associated with this gene.

View Full Product Details