Product Details
- SNP ID
-
rs430387
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:21406307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCCTTGGGCACCCCTGCCTGGG[C/T]CCCCTGGCTGCCCCTTGGCTCCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VENTXP7
PubMed Links
Gene Details
- Gene
- VENTXP7
- Gene Name
- VENT homeobox pseudogene 7
There are no transcripts associated with this gene.
View Full Product Details