Product Details

SNP ID
rs2277748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1495635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCCTGCCAGGGCGGGCACAGCC[A/G]TGGAGCGCATGGGGCTTGGGGATTC
Phenotype
MIM: 600487 MIM: 609346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL5 PubMed Links

Gene Details

Gene
ADAMTSL5
Gene Name
ADAMTS like 5
There are no transcripts associated with this gene.

Gene
PCSK4
Gene Name
proprotein convertase subtilisin/kexin type 4
There are no transcripts associated with this gene.

Gene
REEP6
Gene Name
receptor accessory protein 6
There are no transcripts associated with this gene.

View Full Product Details