Product Details
- SNP ID
-
rs2644112
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:201836978 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTATATATCTGCTTTTGAGCAAATA[C/T]ATGTTACTTGAACTTTAATGCTGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IPO9
PubMed Links
Gene Details
- Gene
- IPO9
- Gene Name
- importin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018085.4 |
|
Intron |
|
|
NP_060555.2 |
- Gene
- IPO9-AS1
- Gene Name
- IPO9 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details