Product Details

SNP ID
rs2820295
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:201831740 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAACATAGAACCCAGCTTTCCA[A/G]CCGGGATTTAGCCAACTATCTGGCC
Phenotype
MIM: 611628
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPO9 PubMed Links

Gene Details

Gene
IPO9
Gene Name
importin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018085.4 Intron NP_060555.2
Gene
IPO9-AS1
Gene Name
IPO9 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NAV1
Gene Name
neuron navigator 1
There are no transcripts associated with this gene.

View Full Product Details