Product Details
- SNP ID
-
rs12538049
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:55812756 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAAGAAAAGAGGCACTGACGATTA[C/T]AGTAAAGAGAGTCTTGCATAGCCTA
- Phenotype
-
MIM: 612140
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEPT14
PubMed Links
Gene Details
- Gene
- SEPT14
- Gene Name
- septin 14
View Full Product Details