Product Details

SNP ID
rs11650527
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3637914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCCTGTCTCCAGTGCACACAGC[A/C]CTTGCCACTATTTGTGTACCCTCTC
Phenotype
MIM: 606272 MIM: 605060
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links
Additional Information
For this assay, SNP(s) [rs59279689] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031681.2 Intron NP_001026851.2
NM_004937.2 Intron NP_004928.2
XM_005256485.2 Intron XP_005256542.1
XM_006721463.2 Intron XP_006721526.1
XM_006721464.2 Intron XP_006721527.1
XM_011523691.2 Intron XP_011521993.1
XM_011523692.2 Intron XP_011521994.1
XM_017024254.1 Intron XP_016879743.1
XM_017024255.1 Intron XP_016879744.1
XM_017024256.1 Intron XP_016879745.1
XM_017024257.1 Intron XP_016879746.1
XM_017024258.1 Intron XP_016879747.1
Gene
SHPK
Gene Name
sedoheptulokinase
There are no transcripts associated with this gene.

View Full Product Details