Product Details

SNP ID
rs11156600
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:155211883 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGTTATGGCTCATGTCTCAAACCC[C/T]TGAGTTTCTTATTCTGTGCCATATC
Phenotype
MIM: 300133
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VBP1 PubMed Links

Gene Details

Gene
VBP1
Gene Name
VHL binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303543.1 Intron NP_001290472.1
NM_001303544.1 Intron NP_001290473.1
NM_001303545.1 Intron NP_001290474.1
NM_003372.6 Intron NP_003363.1

View Full Product Details