Product Details

SNP ID
rs10852753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74539870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGGGTTTCGTCTGGACTGGTAG[C/T]TCATGGAGAACCAGGGCAGTATCTT
Phenotype
MIM: 606786 MIM: 610705
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD300C PubMed Links

Gene Details

Gene
CD300C
Gene Name
CD300c molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006678.4 Intron NP_006669.1
XM_017024033.1 Intron XP_016879522.1
XM_017024034.1 Intron XP_016879523.1
Gene
CD300LB
Gene Name
CD300 molecule like family member b
There are no transcripts associated with this gene.

View Full Product Details