Product Details

SNP ID
rs3087732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8783827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACAGCTGAAGAGTCCTGTAGAG[C/G]AGAGTGATTTTTGTATCTCCAACCC
Phenotype
MIM: 605352
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MFHAS1 PubMed Links

Gene Details

Gene
MFHAS1
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004225.2 Intron NP_004216.2
XM_011543852.2 Intron XP_011542154.1

View Full Product Details