Product Details

SNP ID
rs34141396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70273648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGAAAGATTGATAAGCCAAA[C/T]GTGGGTTTCCTTTTTAGCAACAATA
Phenotype
MIM: 611054
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPFIA1 PubMed Links

Gene Details

Gene
PPFIA1
Gene Name
PTPRF interacting protein alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003626.3 Intron NP_003617.1
NM_177423.2 Intron NP_803172.1
XM_006718716.3 Intron XP_006718779.1
XM_011545306.2 Intron XP_011543608.1
XM_011545307.2 Intron XP_011543609.1
XM_011545308.2 Intron XP_011543610.1
XM_011545309.2 Intron XP_011543611.1
XM_011545310.2 Intron XP_011543612.1
XM_011545311.2 Intron XP_011543613.1
XM_011545312.2 Intron XP_011543614.1
XM_011545313.2 Intron XP_011543615.1
XM_011545314.2 Intron XP_011543616.1
XM_011545315.2 Intron XP_011543617.1
XM_011545316.2 Intron XP_011543618.1
XM_011545317.2 Intron XP_011543619.1
XM_011545318.2 Intron XP_011543620.1
XM_011545319.1 Intron XP_011543621.1
XM_017018448.1 Intron XP_016873937.1
XM_017018449.1 Intron XP_016873938.1
XM_017018450.1 Intron XP_016873939.1

View Full Product Details