Product Details

SNP ID
rs17059945
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:76825201 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTGTGGCTCCAAGTGATTTAGCGT[C/G]TTCAGAGAGGCCTTCCTCAAACCTG
Phenotype
MIM: 604760
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100131655 PubMed Links

Gene Details

Gene
LOC100131655
Gene Name
uncharacterized LOC100131655
There are no transcripts associated with this gene.

Gene
ZNF236
Gene Name
zinc finger protein 236
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306089.1 Intron NP_001293018.1
NM_007345.3 Intron NP_031371.3
XM_005266762.3 Intron XP_005266819.1
XM_011526165.2 Intron XP_011524467.1
XM_011526166.2 Intron XP_011524468.1
XM_011526168.2 Intron XP_011524470.1
XM_011526169.2 Intron XP_011524471.1

View Full Product Details