Product Details
- SNP ID
-
rs9309638
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:86749225 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAAAATAATTTACTTAAAAATACT[A/G]ATGAACTGTTATAAAATTAGCTCTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RMND5A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116452736] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RMND5A
- Gene Name
- required for meiotic nuclear division 5 homolog A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022780.3 |
|
Intron |
|
|
NP_073617.1 |
View Full Product Details