Product Details
- SNP ID
-
rs1011041
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99257341 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGCTATGAAACCATATTAGTACTTT[G/T]TAACCCTTTAACGAGAAGTGGTATA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HSP90B2P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75540530] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSP90B2P
- Gene Name
- heat shock protein 90 beta family member 2, pseudogene
There are no transcripts associated with this gene.
- Gene
- LRRC28
- Gene Name
- leucine rich repeat containing 28
- Gene
- TTC23
- Gene Name
- tetratricopeptide repeat domain 23
There are no transcripts associated with this gene.
View Full Product Details