Product Details

SNP ID
rs1983191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:1009833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTAGTCTCTGTACATCGCCTTCC[A/G]TGCTTTAGACAGGACATCTGCCACA
Phenotype
MIM: 604935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMRT2 PubMed Links

Gene Details

Gene
DMRT2
Gene Name
doublesex and mab-3 related transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130865.2 Intron NP_001124337.1
NM_006557.6 Intron NP_006548.1
NM_181872.4 Intron NP_870987.2
XM_011517687.1 Intron XP_011515989.1
XM_011517690.2 Intron XP_011515992.1
XM_011517694.2 Intron XP_011515996.1
XM_017014213.1 Intron XP_016869702.1
XM_017014214.1 Intron XP_016869703.1
XM_017014215.1 Intron XP_016869704.1
XM_017014216.1 Intron XP_016869705.1

View Full Product Details