Product Details

SNP ID
rs12422183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3634849 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGACAATGGTGGGGAGAGAGGAAA[C/G]GGGGGACAGGCAGGAGAAATGCGGG
Phenotype
MIM: 610625
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ART5 PubMed Links

Gene Details

Gene
ART5
Gene Name
ADP-ribosyltransferase 5
There are no transcripts associated with this gene.

Gene
TRPC2
Gene Name
transient receptor potential cation channel subfamily C member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details