Product Details

SNP ID
rs12440150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84658375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGAGACCTGCACACCCACACT[C/T]TGGCCCGCACGCGGGCACCGCTTCC
Phenotype
MIM: 162340 MIM: 616144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMB PubMed Links

Gene Details

Gene
NMB
Gene Name
neuromedin B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021077.3 197 UTR 5 NP_066563.2
NM_205858.1 197 UTR 5 NP_995580.1
XM_017022239.1 197 UTR 5 XP_016877728.1
Gene
WDR73
Gene Name
WD repeat domain 73
There are no transcripts associated with this gene.

View Full Product Details