Product Details

SNP ID
rs11713327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132418415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTCAGTGAAGTGGCTGTTAATC[A/G]CTTGATACTTAACTATGTTTAAGAG
Phenotype
MIM: 614334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC13 PubMed Links

Gene Details

Gene
DNAJC13
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015268.3 Intron NP_056083.3
XM_005247245.4 Intron XP_005247302.1
XM_017006036.1 Intron XP_016861525.1

View Full Product Details