Product Details

SNP ID
rs12778953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87054855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCATCCAATGCCCACAACAAGAA[A/C]TGGACCTAAGAAATGAAAGCACAAA
Phenotype
MIM: 138130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLUD1 PubMed Links

Gene Details

Gene
GLUD1
Gene Name
glutamate dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318900.1 Intron NP_001305829.1
NM_001318901.1 Intron NP_001305830.1
NM_001318902.1 Intron NP_001305831.1
NM_001318904.1 Intron NP_001305833.1
NM_001318905.1 Intron NP_001305834.1
NM_001318906.1 Intron NP_001305835.1
NM_005271.4 Intron NP_005262.1

View Full Product Details