Product Details

SNP ID
rs2281748
Assay Type
Validated
NCBI dbSNP Submissions
69
Location
Chr.1:19074911 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TGATAAAAGACCTGCAAAGCACAAC[A/G]GGCAGAGGTGTGTCAGGAGCAGGCA
Phenotype
MIM: 609890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBR4 PubMed Links

Gene Details

Gene
UBR4
Gene Name
ubiquitin protein ligase E3 component n-recognin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020765.2 Intron NP_065816.2
XM_011541108.2 Intron XP_011539410.1
XM_011541109.2 Intron XP_011539411.1
XM_011541110.2 Intron XP_011539412.1
XM_011541111.2 Intron XP_011539413.1
XM_011541112.2 Intron XP_011539414.1
XM_011541113.2 Intron XP_011539415.1
XM_011541114.2 Intron XP_011539416.1
XM_011541115.2 Intron XP_011539417.1
XM_011541116.2 Intron XP_011539418.1
XM_011541117.2 Intron XP_011539419.1
XM_011541118.2 Intron XP_011539420.1
XM_011541119.2 Intron XP_011539421.1
XM_011541120.2 Intron XP_011539422.1
XM_011541121.2 Intron XP_011539423.1
XM_017000822.1 Intron XP_016856311.1
XM_017000823.1 Intron XP_016856312.1
XM_017000824.1 Intron XP_016856313.1
XM_017000825.1 Intron XP_016856314.1
XM_017000826.1 Intron XP_016856315.1
XM_017000827.1 Intron XP_016856316.1
XM_017000828.1 Intron XP_016856317.1
XM_017000829.1 Intron XP_016856318.1
XM_017000830.1 Intron XP_016856319.1

View Full Product Details