Product Details

SNP ID
rs315125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69365104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTTGGGACGTGTATTTCTATA[C/T]ATTACAAATCATAAAAGAATTCAAC
Phenotype
MIM: 602116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YEATS4 PubMed Links

Gene Details

Gene
YEATS4
Gene Name
YEATS domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300950.1 Intron NP_001287879.1
NM_006530.3 Intron NP_006521.1

View Full Product Details