Product Details
- SNP ID
-
rs11591216
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
22
- Location
-
Chr.1:26534353 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGGACAAGCCATGGCCCCTTCC[C/T]GAGTTAACCTGCAGCATGCTTAGGT
- Phenotype
-
MIM: 601684
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RPS6KA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77010086] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPS6KA1
- Gene Name
- ribosomal protein S6 kinase A1
View Full Product Details