Product Details

SNP ID
rs6752257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172560527 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAAGGTTAAATCTTTTGCGTAAG[A/G]CAGTACCATTGATAGTGGAGTTGGG
Phenotype
MIM: 602524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDK1 PubMed Links

Gene Details

Gene
PDK1
Gene Name
pyruvate dehydrogenase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278549.1 Intron NP_001265478.1
NM_002610.4 Intron NP_002601.1
XM_006712594.2 Intron XP_006712657.1
XM_006712595.3 Intron XP_006712658.1
XM_011511343.2 Intron XP_011509645.1
XM_011511344.2 Intron XP_011509646.1
XM_011511345.2 Intron XP_011509647.1
XM_011511347.2 Intron XP_011509649.1

View Full Product Details