Product Details

SNP ID
rs908695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36674655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGTGGAGGGTCATTCTTGGGT[G/T]TGTTTTGCTCCTGTCTGATTTTCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERTM1 PubMed Links

Gene Details

Gene
SERTM1
Gene Name
serine rich and transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203451.2 Intron NP_982276.2

View Full Product Details