Product Details

SNP ID
rs1058751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92210634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAAGTGTGAAGATTACTGTGAGG[A/T]CTCAAGTTACTTGACTAATCAATCC
Phenotype
MIM: 600709
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IARS PubMed Links

Gene Details

Gene
IARS
Gene Name
isoleucyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002161.5 4285 UTR 3 NP_002152.2
NM_013417.3 4285 UTR 3 NP_038203.2

View Full Product Details