Product Details

SNP ID
rs647065
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:84400671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAGCTCTCAACTGGTGTGCCAC[A/G]GCACACTGATAGTTCACAAACCCTT
Phenotype
MIM: 608057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNASE2B PubMed Links

Gene Details

Gene
DNASE2B
Gene Name
deoxyribonuclease II beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021233.2 Intron NP_067056.2
NM_058248.1 Intron NP_490649.1
XM_011541878.2 Intron XP_011540180.1

View Full Product Details