Product Details

SNP ID
rs34427967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36750565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGGATGAATCCGATGATGCA[C/G]AGTGAGAGATGTCTGTAGGCAGAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 538 Missense Mutation CTG,GTG L159V NP_001180481.1
NM_001267779.1 538 Missense Mutation CTG,GTG L127V NP_001254708.1

View Full Product Details