Product Details

SNP ID
rs11620864
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52432953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTAAAACGTTAAATGAAAAAG[C/T]CACCTGCTTCCCTCTCTACTGCCAA
Phenotype
MIM: 616179
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TXNDC16 PubMed Links

Gene Details

Gene
TXNDC16
Gene Name
thioredoxin domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160047.1 Intron NP_001153519.1
NM_020784.2 Intron NP_065835.2
XM_017021505.1 Intron XP_016876994.1

View Full Product Details