Product Details
- SNP ID
-
rs11813262
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:110926634 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAACTTAATGTTATGACAAACATG[C/T]GTAGAATCTTTCAAGACACTTTGTT
- Phenotype
-
MIM: 613605
MIM: 602775
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BBIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs371275958] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BBIP1
- Gene Name
- BBSome interacting protein 1
There are no transcripts associated with this gene.
- Gene
- RPL13AP6
- Gene Name
- ribosomal protein L13a pseudogene 6
There are no transcripts associated with this gene.
- Gene
- SHOC2
- Gene Name
- SHOC2, leucine rich repeat scaffold protein
View Full Product Details