Product Details

SNP ID
rs912075
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:42734855 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGGACCTCTGAATTATTTCTTGCT[C/T]AGCCCTGGATGTGCTATGTAACCCA
Phenotype
MIM: 610036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN19 PubMed Links

Gene Details

Gene
CLDN19
Gene Name
claudin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123395.1 1097 UTR 3 NP_001116867.1
NM_001185117.1 1097 UTR 3 NP_001172046.1
NM_148960.2 1097 UTR 3 NP_683763.2

View Full Product Details