Product Details

SNP ID
rs2199161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72114529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTTCCTGGCAATTGAGAATGCT[C/G]TATACAAAGGTACAATGATGGATAC
Phenotype
MIM: 157129
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP1B PubMed Links

Gene Details

Gene
MAP1B
Gene Name
microtubule associated protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324255.1 Intron NP_001311184.1
NM_005909.4 Intron NP_005900.2

View Full Product Details