Product Details

SNP ID
rs2069910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127420398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACTCAAATGCACACTGGCCTCA[C/T]GGCTGCCCTGCCCCAACCCCTTTCC
Phenotype
MIM: 612283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4783 PubMed Links

Gene Details

Gene
MIR4783
Gene Name
microRNA 4783
There are no transcripts associated with this gene.

Gene
PROC
Gene Name
protein C, inactivator of coagulation factors Va and VIIIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000312.3 Intron NP_000303.1
XM_005263717.4 Intron XP_005263774.1
XM_017004505.1 Intron XP_016859994.1
XM_017004506.1 Intron XP_016859995.1

View Full Product Details